Learn More
Abcam Luteolin, Antio x idant and anti-inflammatory, 10MG
Catalog No. 502707372
Encompass_Preferred
MW 286.24 g/mol, Purity >99%. Antioxidant, anti-inflammatory and anticancer flavonoid. Free radical scavenger and apoptosis inducer. Inhibits TNF-α induced COX-2 expression and supresses PI3K/Akt, NF-κB cell survival pathways.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7372 Supplier Abcam Supplier No. AB12066210MG
MW 286.24 g/mol, Purity >99%. Antioxidant, anti-inflammatory and anticancer flavonoid. Free radical scavenger and apoptosis inducer. Inhibits TNF-α induced COX-2 expression and supresses PI3K/Akt, NF-κB cell survival pathways.
The product is subject to the following: Abcam Restricted Use Statement