Learn More
Cayman Chemical Luteolin, Purity: ≥98%, CAS: 491-70-3, 500 mg
SDP
Catalog No. 502888763
Encompass_Preferred
A common dietary phenolic flavone; inhibits soybean and reticulocyte 15-lipoxygenases (IC50 = 0.6 µM); inhibits the release of TNFα from neutrophils; inhibits matrix metalloproteinases
Catalog No. 50-288-8763 Supplier Cayman Chemical Supplier No. 10004161500 MG
A common dietary phenolic flavone; inhibits soybean and reticulocyte 15-lipoxygenases (IC50 = 0.6 µM); inhibits the release of TNFα from neutrophils; inhibits matrix metalloproteinases