Learn More
Medchemexpress LLC Luteolin monohydrate | 6113-16-2 | 99.1% | 304.25 | C15H12O7 | 100 MG
SDP
Catalog No. 5000282169
Encompass_Preferred
Luteolin monohydrate is the monohydrate form of luteolin, a naturally occurring flavonoid used in biological research. It acts as a potent Nrf2 inhibitor and has reported anti-inflammatory and anticancer activities, including induction of apoptosis and cell-cycle arrest in various cancer cell lines.
  • Monohydrate form suitable for research applications.
  • Potent Nrf2 inhibitor with reported anti-inflammatory activity.
  • Induces apoptosis and cell-cycle arrest in multiple cancer cell lines.
  • High purity (99.13%).
  • Chemical identifiers: CAS 6113-16-2; molecular weight 304.25; formula C15H12O7.
  • Available as solid and as a DMSO solution format.
Catalog No. 50-002-82169 Supplier Medchemexpress LLC Supplier No. HYW303895100MG
Luteolin monohydrate is the monohydrate form of luteolin, a naturally occurring flavonoid used in biological research. It acts as a potent Nrf2 inhibitor and has reported anti-inflammatory and anticancer activities, including induction of apoptosis and cell-cycle arrest in various cancer cell lines.
  • Monohydrate form suitable for research applications.
  • Potent Nrf2 inhibitor with reported anti-inflammatory activity.
  • Induces apoptosis and cell-cycle arrest in multiple cancer cell lines.
  • High purity (99.13%).
  • Chemical identifiers: CAS 6113-16-2; molecular weight 304.25; formula C15H12O7.
  • Available as solid and as a DMSO solution format.