Learn More
Medchemexpress LLC Luteolin monohydrate | 6113-16-2 | 99.1% | 304.25 | C15H12O7 | 500 MG
SDP
Catalog No. 5000282518
Encompass_Preferred
Luteolin monohydrate is the monohydrate form of luteolin, a naturally occurring flavonoid used in biochemical and pharmacological research. It is reported to act as an Nrf2 inhibitor and has documented anti-inflammatory and anticancer activities. Supplied as a solid with high reported purity for laboratory studies.
  • High purity: 99.1%.
  • Flavonoid with reported anti-inflammatory and anticancer activity.
  • Reported Nrf2 inhibitor activity.
  • Molecular formula C15H12O7; molecular weight 304.25.
  • Available in a 500 mg pack for laboratory use.
Catalog No. 50-002-82518 Supplier Medchemexpress LLC Supplier No. HYW303895500MG
Luteolin monohydrate is the monohydrate form of luteolin, a naturally occurring flavonoid used in biochemical and pharmacological research. It is reported to act as an Nrf2 inhibitor and has documented anti-inflammatory and anticancer activities. Supplied as a solid with high reported purity for laboratory studies.
  • High purity: 99.1%.
  • Flavonoid with reported anti-inflammatory and anticancer activity.
  • Reported Nrf2 inhibitor activity.
  • Molecular formula C15H12O7; molecular weight 304.25.
  • Available in a 500 mg pack for laboratory use.