Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Luteolin monohydrate | 6113-16-2 | 99.1% | 304.25 g/mol | C15H12O7 | 1 ML
SDP

Catalog No. 5000281966
Encompass_Preferred

Luteolin monohydrate is the monohydrate form of luteolin, a flavonoid research reagent reported to have anti-inflammatory and anticancer activity. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and as solid quantities for laboratory use, and is intended for research applications.

  • Ready-to-use 10 mM solution in DMSO for convenient dosing
  • High purity (99.1%) for reproducible results
  • Suitable for cell-based and biochemical assays
  • Solid forms available for bulk use (100 MG-5 G)
  • Molecular formula C15H12O7 and molecular weight 304.25 g/mol
  • Storage recommendations provided for solid and solution forms

Catalog No. 50-002-81966 Supplier Medchemexpress LLC Supplier No. HYW30389510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Luteolin monohydrate is the monohydrate form of luteolin, a flavonoid research reagent reported to have anti-inflammatory and anticancer activity. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and as solid quantities for laboratory use, and is intended for research applications.

  • Ready-to-use 10 mM solution in DMSO for convenient dosing
  • High purity (99.1%) for reproducible results
  • Suitable for cell-based and biochemical assays
  • Solid forms available for bulk use (100 MG-5 G)
  • Molecular formula C15H12O7 and molecular weight 304.25 g/mol
  • Storage recommendations provided for solid and solution forms

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.