Learn More
Medchemexpress LLC Luteolin monohydrate | 6113-16-2 | MFCD00017309 | 99.1% | 304.25 g/mol | C15H12O7 | 250 MG
SDP
Catalog No. 5000281954
Encompass_Preferred
Luteolin monohydrate is the monohydrate form of luteolin, a naturally occurring flavonoid used in life-science research. It is reported to act as an Nrf2 pathway modulator and shows anti-inflammatory and anticancer activity in cellular studies. It is supplied as a solid and provided in small research quantities for biochemical assays and mechanistic investigations.
  • Monohydrate form of luteolin
  • Reported Nrf2 inhibitor and bioactive flavonoid
  • High purity (99.13%)
  • Available in small research quantities, e.g., 250 mg
  • Storage: 4°C, sealed, away from moisture; in solution: -80°C (6 months)
Catalog No. 50-002-81954 Supplier Medchemexpress LLC Supplier No. HYW303895250MG
Luteolin monohydrate is the monohydrate form of luteolin, a naturally occurring flavonoid used in life-science research. It is reported to act as an Nrf2 pathway modulator and shows anti-inflammatory and anticancer activity in cellular studies. It is supplied as a solid and provided in small research quantities for biochemical assays and mechanistic investigations.
  • Monohydrate form of luteolin
  • Reported Nrf2 inhibitor and bioactive flavonoid
  • High purity (99.13%)
  • Available in small research quantities, e.g., 250 mg
  • Storage: 4°C, sealed, away from moisture; in solution: -80°C (6 months)