Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Luteolin, MP Biomedicals

Catalog No. ICN223887 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN223887 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN223887 Supplier MP Biomedicals, Inc Supplier No. 0522388750

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 491-70-3
Molecular Formula C15H10O6
Molecular Weight (g/mol) 286.239
InChI Key IQPNAANSBPBGFQ-UHFFFAOYSA-N
Synonym luteolin, 3',4',5,7-tetrahydroxyflavone, 2-3,4-dihydroxyphenyl-5,7-dihydroxy-4h-chromen-4-one, digitoflavone, luteolol, flacitran, luteoline, weld lake, cyanidenon 1470, salifazide
PubChem CID 5280445
ChEBI CHEBI:15864
IUPAC Name 2-(3,4-dihydroxyphenyl)-5,7-dihydroxychromen-4-one
SMILES C1=CC(=C(C=C1C2=CC(=O)C3=C(C=C(C=C3O2)O)O)O)O

Specifications

Quantity 100 mg
Formula Weight 286
Chemical Name or Material Luteolin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.