Learn More
Medchemexpress LLC Luteolinidin chloride | 1154-78-5 | MFCD00016779 | 99.7% | C15H11ClO5 | 10MG
SDP
Catalog No. 5000260904
Encompass_Preferred
Luteolinidin chloride is an anthocyanidin chloride research compound used in biochemical studies. It functions as a CD38 inhibitor (Ki = 11.4 μM) and a competitive tyrosinase inhibitor (IC50 = 3.7 μM), and has reported cardioprotective effects in ischemia/reperfusion models via preservation of endothelial nitric oxide synthase. The material is supplied as a brown to reddish-brown solid with high purity.
Catalog No. 50-002-60904 Supplier Medchemexpress LLC Supplier No. HY12999710MG
Luteolinidin chloride is an anthocyanidin chloride research compound used in biochemical studies. It functions as a CD38 inhibitor (Ki = 11.4 μM) and a competitive tyrosinase inhibitor (IC50 = 3.7 μM), and has reported cardioprotective effects in ischemia/reperfusion models via preservation of endothelial nitric oxide synthase. The material is supplied as a brown to reddish-brown solid with high purity.