Learn More
Cayman Chemical Luteoloside, Purity: ≥98%, CAS: 5373-11-5, 10 mg
SDP
Catalog No. 502935214
Encompass_Preferred
A flavanoid with diverse biological activities; reduces LDH and caspase-3 activities and production of ROS and increases cell viability and 14-3-3η protein levels in H9C2 cardiomyocytes after anoxia/reoxygenation injury; blocks 3C protease activity (IC50 = 0.36 mM); reduces the cytopathic effect of enterovirus 71 in rhabdomyosarcoma cells (EC50 = 0.43 mM); inhibits the growth of HCC cells in vitro and reduces the number of lung metastases in an SMMC-7721 HCC mouse xenograft model at 2 mg/kg; reduces acanthosis and expression of epidermal differentiation markers in a mouse model of imiquimod-induced psoriasis
Catalog No. 50-293-5214 Supplier Cayman Chemical Supplier No. 2514110 MG
A flavanoid with diverse biological activities; reduces LDH and caspase-3 activities and production of ROS and increases cell viability and 14-3-3η protein levels in H9C2 cardiomyocytes after anoxia/reoxygenation injury; blocks 3C protease activity (IC50 = 0.36 mM); reduces the cytopathic effect of enterovirus 71 in rhabdomyosarcoma cells (EC50 = 0.43 mM); inhibits the growth of HCC cells in vitro and reduces the number of lung metastases in an SMMC-7721 HCC mouse xenograft model at 2 mg/kg; reduces acanthosis and expression of epidermal differentiation markers in a mouse model of imiquimod-induced psoriasis