Learn More
Cayman Chemical LuteoretIculIn 2.5mg
SDP
Catalog No. 503015700
Encompass_Preferred
A nitro-containing bacterial metabolite; has mosquitocidal activity against A. aegypti at 6.25 mg/L; has nematocidal activity against C. elegans
Catalog No. 50-301-5700 Supplier Cayman Chemical Supplier No. 255152.5 MG
A nitro-containing bacterial metabolite; has mosquitocidal activity against A. aegypti at 6.25 mg/L; has nematocidal activity against C. elegans