Learn More
AdipoGen Luteoreticulin
SDP
Supplier:  AdipoGen BVT0457M001
Encompass_Preferred
Chemical. CAS 22388-89-2. Formula C19H19NO5. MW 341.4. Isolated from Streptomyces thioluteus. Nitrophenyl pyranone derivative related to aureothin Prod. No. BVT-0303 http//www.adipogen.com/bvt-0303/aureothin.html . Insecticidal and nematocidal compound. Metabolites within this product class show pronounced antitumor and immunosuppressive activity.
Catalog No. 50-256-1903