Learn More
Cayman Chemical Thiolutin, Purity: ≥97%, CAS: 87-11-6, 10 mg
SDP
Catalog No. 502951294
Encompass_Preferred
A natural dithiol that reversibly inhibits bacterial and yeast RNA polymerases (IC50 = 3 μg/ml); used for the analysis of mRNA stability; inhibits endothelial cell adhesion (IC50 induced angiogenesis in mice by inhibiting Hsp27 interactions with cytoskeletal elements
Catalog No. 50-295-1294 Supplier Cayman Chemical Supplier No. 1135010 MG
A natural dithiol that reversibly inhibits bacterial and yeast RNA polymerases (IC50 = 3 μg/ml); used for the analysis of mRNA stability; inhibits endothelial cell adhesion (IC50 induced angiogenesis in mice by inhibiting Hsp27 interactions with cytoskeletal elements