Learn More
Strem, An Ascensus Company CAS# 10099-66-8. 5g. Lutetium(III) chloride, anhydrous (99.9%-Lu) (REO). MFCD00011099
SDP
Catalog No. 5090116241
Encompass
CAS# 10099-66-8. 5g. Lutetium(III) chloride, anhydrous (99.9%-Lu) (REO). MFCD00011099. Molecular Weight: 281.33. Molecular Formula: LuCl3. Color/form: white pwdr. Strem# 93-7101. http://www.strem.com/catalog/v/93-7101/
Catalog No. 50-901-16241 Supplier Strem, An Ascensus Company Supplier No. 9371015G
CAS# 10099-66-8. 5g. Lutetium(III) chloride, anhydrous (99.9%-Lu) (REO). MFCD00011099. Molecular Weight: 281.33. Molecular Formula: LuCl3. Color/form: white pwdr. Strem# 93-7101. http://www.strem.com/catalog/v/93-7101/