Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lutetium(III) chloride, ultra dry, 99.98% (REO)

Catalog No. AA3580206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3580206 5 g
AA3580203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3580206 Supplier Thermo Scientific Chemicals Supplier No. 03580206
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10099-66-8

It is used as an additive for chemical industry, single crystal scintillators, carbon arc rods, PET scan detectors, high refractive index glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10099-66-8
MDL Number MFCD00011099

Specifications

Melting Point 905°C (sublimation)
Physical Form Powder
Quantity 5 g
Packaging Ampule
Sensitivity Hygroscopic
Density 3.98 g/mL
Merck Index 14,5615
Solubility Information Soluble in water.
Formula Weight 281.33
Percent Purity 99.98%
Assay (REO)
Chemical Name or Material Lutetium(III) chloride, Ultra dry
Show More Show Less
Hazard Category H315-H319-H335-H500
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-240-1
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.