Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lutetium(III) oxide, REacton™, 99.995% (metals basis), REM 50ppm

Catalog No. AA1091203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1091203 1 g
AA1091206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1091203 Supplier Thermo Scientific Chemicals Supplier No. 01091203
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 12032-20-1 | Lu2O3 | 397.93 g/mol

The product of Lutetium(III) oxide and palladium(II) oxide has a potential use in superconducting materials. It is an important raw material for laser crystals. It has specialized uses in ceramics, glass, phosphors, and lasers. Also used as a catalyst in cracking, alkylation, hydrogenation, and polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12032-20-1
Molecular Formula Lu2O3
Molecular Weight (g/mol) 397.93
MDL Number MFCD00011100
InChI Key UGBIHFMRUDAMBY-UHFFFAOYSA-N
Synonym lutetium 3+ ; oxygen 2-, lutetium oxide, lutetium iii oxide trace rare earth metals basis 1g, cid159406, einecs 234-764-3, lu2o3, lutetium 3+
IUPAC Name dilutetium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Lu+3].[Lu+3]

Specifications

Melting Point 2487°C
Physical Form Powder
Quantity 1 g
Merck Index 14,5615
Solubility Information Insoluble in water.
Formula Weight 397.94
Percent Purity 99.995%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Density 9.420 g/mL
Assay (metals basis)
Chemical Name or Material Lutetium(III) oxide, REM 50ppm
Show More Show Less
Hazard Category H315-H320-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 234-764-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.