Learn More
Medchemexpress LLC Luvixasertib hydrochloride | 1610677-37-6 | 99.6% | 535.04 | C28H31ClN6O3 | 10 MG
SDP
Catalog No. 5000211427
Encompass_Preferred
Luvixasertib hydrochloride is the hydrochloride salt of luvixasertib, a highly selective and orally bioavailable inhibitor of the TTK/Mps1 protein kinase. It is used in preclinical cancer research to probe spindle assembly checkpoint biology and to evaluate therapeutic potential against TTK-dependent tumors; reported enzymatic potency is approximately IC50 1.7 nM.
  • Highly potent TTK/Mps1 inhibition (IC50 ≈ 1.7 nM).
  • Orally bioavailable small-molecule inhibitor suitable for in vivo and in vitro studies.
  • Hydrochloride salt form for improved solubility and handling.
  • High purity solid for research use (purity 99.6%).
  • Analytical identifiers provided: CAS number 1610677-37-6; molecular weight 535.04 g/mol.
Catalog No. 50-002-11427 Supplier Medchemexpress LLC Supplier No. HY101340A10MG
Luvixasertib hydrochloride is the hydrochloride salt of luvixasertib, a highly selective and orally bioavailable inhibitor of the TTK/Mps1 protein kinase. It is used in preclinical cancer research to probe spindle assembly checkpoint biology and to evaluate therapeutic potential against TTK-dependent tumors; reported enzymatic potency is approximately IC50 1.7 nM.
  • Highly potent TTK/Mps1 inhibition (IC50 ≈ 1.7 nM).
  • Orally bioavailable small-molecule inhibitor suitable for in vivo and in vitro studies.
  • Hydrochloride salt form for improved solubility and handling.
  • High purity solid for research use (purity 99.6%).
  • Analytical identifiers provided: CAS number 1610677-37-6; molecular weight 535.04 g/mol.