Learn More
Medchemexpress LLC Luvixasertib hydrochloride | 1610677-37-6 | 99.6% | 535.04 g/mol | C28H31ClN6O3 | 25 MG
SDP
Catalog No. 5000211990
Encompass_Preferred
Luvixasertib hydrochloride is a selective, orally bioavailable TTK/Mps1 kinase inhibitor used in preclinical research to probe mitotic checkpoint function and to evaluate anticancer activity. It demonstrates potent in vitro inhibition of TTK and is supplied as a high-purity hydrochloride solid suitable for laboratory studies.
  • Selective TTK/Mps1 kinase inhibition for targeted research.
  • Potent in vitro activity (IC50 1.7 nM) for sensitive assays.
  • Orally bioavailable profile applicable to in vivo models.
  • Supplied as a hydrochloride solid with high purity for reproducible results.
  • Stable when stored according to recommended conditions for research use.
Catalog No. 50-002-11990 Supplier Medchemexpress LLC Supplier No. HY101340A25MG
Luvixasertib hydrochloride is a selective, orally bioavailable TTK/Mps1 kinase inhibitor used in preclinical research to probe mitotic checkpoint function and to evaluate anticancer activity. It demonstrates potent in vitro inhibition of TTK and is supplied as a high-purity hydrochloride solid suitable for laboratory studies.
  • Selective TTK/Mps1 kinase inhibition for targeted research.
  • Potent in vitro activity (IC50 1.7 nM) for sensitive assays.
  • Orally bioavailable profile applicable to in vivo models.
  • Supplied as a hydrochloride solid with high purity for reproducible results.
  • Stable when stored according to recommended conditions for research use.