Learn More
Medchemexpress LLC Luvixasertib hydrochloride | 1610677-37-6 | 99.6% | 535.04 g/mol | C28H31ClN6O3 | 50 MG
SDP
Catalog No. 5000211761
Encompass_Preferred
Luvixasertib hydrochloride is a selective, orally bioavailable small-molecule inhibitor of TTK/Mps1 used for preclinical research. It inhibits TTK/Mps1 with an in vitro IC50 of 1.7 nM and has demonstrated anticancer activity in cell-based studies.
  • Selective TTK/Mps1 inhibition with IC50 of 1.7 nM.
  • Orally bioavailable scaffold suitable for in vivo studies.
  • Hydrochloride salt form improves solubility and handling.
  • High reported purity (≈99.6%) for research reproducibility.
  • Supplied in small research quantities for assay development and compound characterization.
Catalog No. 50-002-11761 Supplier Medchemexpress LLC Supplier No. HY101340A50MG
Luvixasertib hydrochloride is a selective, orally bioavailable small-molecule inhibitor of TTK/Mps1 used for preclinical research. It inhibits TTK/Mps1 with an in vitro IC50 of 1.7 nM and has demonstrated anticancer activity in cell-based studies.
  • Selective TTK/Mps1 inhibition with IC50 of 1.7 nM.
  • Orally bioavailable scaffold suitable for in vivo studies.
  • Hydrochloride salt form improves solubility and handling.
  • High reported purity (≈99.6%) for research reproducibility.
  • Supplied in small research quantities for assay development and compound characterization.