Learn More
Biotang Inc Luxury Microscope Slide Boxes, 100 Place, White,12/Pk. Blue
Catalog No. 507514983 Shop All BIOTANG Inc Products
Encompass_Preferred
Luxury Microscope Slide Boxes, 100 place, blue,12/pk. perfect for storage or transportation of standard size slides. Molded, Keep boxes safely on the benchtop for long-term storage, same as HD15994E
Catalog No. 50-751-4983 Supplier Biotang Inc Supplier No. HD15994A9
Luxury Microscope Slide Boxes, 100 place, blue,12/pk. perfect for storage or transportation of standard size slides. Molded, Keep boxes safely on the benchtop for long-term storage, same as HD15994E