Learn More
Medchemexpress LLC Luxeptinib | 1616428-23-9 | 495.43 | 50 MG
SDP
Catalog No. 5000350819
Encompass_Preferred
Luxeptinib (CG-806) is an orally active, reversible, first-in-class, non-covalent, and potent pan-FLT3/pan-BTK inhibitor. It has been shown to induce cell cycle arrest, apoptosis, or autophagy in acute myeloid leukemia cells. Luxeptinib also inhibits cell proliferation and suppresses BCR signaling-induced phosphorylation.
  • Orally active, reversible, first-in-class, non-covalent, and potent pan-FLT3/pan-BTK inhibitor.
  • Induces cell cycle arrest, apoptosis, or autophagy in acute myeloid leukemia cells.
  • Inhibits cell proliferation with an IC50 of 32 nM in MEC-1 CLL cells.
  • Suppresses BCR signaling-induced phosphorylation of BTK, PLCγ2, AKT, ERK1/2, and S6 ribosomal protein in primary chronic lymphocytic leukemia (CLL) cells.
  • Completely inhibits phosphorylation of FLT3 and STAT5 in MV4-11 cells.
Catalog No. 50-003-50819 Supplier Medchemexpress LLC Supplier No. HY13953550MG
Luxeptinib (CG-806) is an orally active, reversible, first-in-class, non-covalent, and potent pan-FLT3/pan-BTK inhibitor. It has been shown to induce cell cycle arrest, apoptosis, or autophagy in acute myeloid leukemia cells. Luxeptinib also inhibits cell proliferation and suppresses BCR signaling-induced phosphorylation.
  • Orally active, reversible, first-in-class, non-covalent, and potent pan-FLT3/pan-BTK inhibitor.
  • Induces cell cycle arrest, apoptosis, or autophagy in acute myeloid leukemia cells.
  • Inhibits cell proliferation with an IC50 of 32 nM in MEC-1 CLL cells.
  • Suppresses BCR signaling-induced phosphorylation of BTK, PLCγ2, AKT, ERK1/2, and S6 ribosomal protein in primary chronic lymphocytic leukemia (CLL) cells.
  • Completely inhibits phosphorylation of FLT3 and STAT5 in MV4-11 cells.