Learn More
Medchemexpress LLC Luxeptinib | 1616428-23-9 | 99.0% | C25H17F4N5O2 | 10MG
SDP
Catalog No. 5000261848
Encompass_Preferred
Luxeptinib is an orally active, reversible, non-covalent small-molecule inhibitor that targets FLT3 and BTK signaling pathways. Provided for research use, it inhibits downstream phosphorylation events (including STAT5, AKT, and ERK1/2), induces cell-cycle arrest and apoptosis in leukemic cells, and includes manufacturer guidance for preparation and storage.
  • Orally active reversible pan-FLT3 and pan-BTK inhibitor.
  • Inhibits phosphorylation of FLT3 and STAT5 in cellular assays.
  • Induces cell-cycle arrest, apoptosis, and autophagy in leukemia models.
  • Supplied as a powder with recommended storage conditions for stability.
  • Manufacturer provides suggested in-vitro stock concentrations and assay guidance.
  • Suitable for in-vitro cell proliferation assays at 0.1-10 μM.
Catalog No. 50-002-61848 Supplier Medchemexpress LLC Supplier No. HY13953510MG
Luxeptinib is an orally active, reversible, non-covalent small-molecule inhibitor that targets FLT3 and BTK signaling pathways. Provided for research use, it inhibits downstream phosphorylation events (including STAT5, AKT, and ERK1/2), induces cell-cycle arrest and apoptosis in leukemic cells, and includes manufacturer guidance for preparation and storage.
  • Orally active reversible pan-FLT3 and pan-BTK inhibitor.
  • Inhibits phosphorylation of FLT3 and STAT5 in cellular assays.
  • Induces cell-cycle arrest, apoptosis, and autophagy in leukemia models.
  • Supplied as a powder with recommended storage conditions for stability.
  • Manufacturer provides suggested in-vitro stock concentrations and assay guidance.
  • Suitable for in-vitro cell proliferation assays at 0.1-10 μM.