Learn More
Diagnostic Biosystems LUXOL FAST BLUE Luxol Fast Blue
Catalog No. 502407548
Encompass_Preferred
The Luxol Fast Blue Stain Kit is designed for staining myelin/myelinated axons and Nissil substance on formalin fixed, paraffin-embedded tissue. This product is used for identifying the basic neuronal structure in brain or spinal cord sections.
Catalog No. 50-240-7548 Supplier Diagnostic Biosystems Supplier No. KT022
The Luxol Fast Blue Stain Kit is designed for staining myelin/myelinated axons and Nissil substance on formalin fixed, paraffin-embedded tissue. This product is used for identifying the basic neuronal structure in brain or spinal cord sections.