Learn More
Newcomer Supply Luxol Fast Blue (LFB) Stain Set, 500 ml set
SDP
Catalog No. NC0795630
Encompass
Solution A: Luxol Fast Blue Stain 0.1%, Alcoholic 500 ml, Solution B: Lithium Carbonate, Saturated Aqueous 500 ml. Newcomer Supply Luxol Fast Blue (LFB) Stain Set, with included microwave modification, is a common procedure for the demonstration of myelin in central nervous system and peripheral nerve tissues. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
Catalog No. NC0795630 Supplier Newcomer Supply Supplier No. 12218A
Solution A: Luxol Fast Blue Stain 0.1%, Alcoholic 500 ml, Solution B: Lithium Carbonate, Saturated Aqueous 500 ml. Newcomer Supply Luxol Fast Blue (LFB) Stain Set, with included microwave modification, is a common procedure for the demonstration of myelin in central nervous system and peripheral nerve tissues. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.