Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Luxol Fast Blue ARN, Thermo Scientific™

Catalog No. AC212160250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC212160250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC212160250 Supplier Thermo Scientific Chemicals Supplier No. 212160250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12226-74-3
Molecular Formula C26H16N3Na3O10S3
Molecular Weight (g/mol) 695.574
InChI Key ADGGJQPKBDIZMT-UHFFFAOYSA-K
Synonym Solvent Blue 37
PubChem CID 19731
IUPAC Name trisodium;4-[(4-anilino-5-sulfonatonaphthalen-1-yl)diazenyl]-5-hydroxynaphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NC2=C3C(=C(C=C2)N=NC4=C5C(=CC(=C4)S(=O)(=O)[O-])C=C(C=C5O)S(=O)(=O)[O-])C=CC=C3S(=O)(=O)[O-].[Na+].[Na+].[Na+]

Specifications

Infrared Spectrum Authentic
Lambda Max 576nm (in methanol)
Formula Weight 695.58
Color Blue/Purple
Quantity 25 g
Physical Form Powder
Chemical Name or Material Luxol(r) Fast Blue ARN
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.