Learn More
Electron Microscopy Sciences Luxol Fast Blue Solution 0.1% in 95% Alcohol 500 ML
SDP
Catalog No. 5032291
Encompass_Preferred
Concentration of 0.1% in 95% alcohol.
Catalog No. 50-322-91 Supplier Electron Microscopy Sciences Supplier No. 2605615
Concentration of 0.1% in 95% alcohol.