Learn More
Luxol™ Fast Blue Solution, 250 mL, G-Biosciences

Description
- This solution can be used with frozen or paraffin-embedded sections, preferably formaldehyde-fixed tissues.
Specifications
Specifications
| Chemical Name or Material | Solvent Blue 38 |
| CAS | 1328-51-4 |
| Color | Blue |
| pH | 3.1 to 5.5 |
| Quantity | 250 mL |
| Molecular Weight (g/mol) | 780.16 |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.