Learn More
Newcomer Supply Luxol Fast Blue (LFB) - Cresyl Violet Stain Kit, 250 ml kit
SDP
Catalog No. NC2102281
Encompass
Newcomer Supply Luxol Fast Blue (LFB) – Cresyl Violet Stain Kit, with included microwave modification, is for the demonstration of myelin and Nissl substance in central nervous system and peripheral nerve tissues.
Catalog No. NC2102281 Supplier Newcomer Supply Supplier No. 9155A
Newcomer Supply Luxol Fast Blue (LFB) – Cresyl Violet Stain Kit, with included microwave modification, is for the demonstration of myelin and Nissl substance in central nervous system and peripheral nerve tissues.