Learn More
Ihc World LLC LUXOL FAST BLUE STAIN KIT
SDP
Supplier:  Ihc World LLC IW3005
Encompass_Preferred
This kit is designed for staining myelin/myelinated axons on formalin-fixed, paraffin-embedded brain and spinal cord tissue sections, as well as frozen sections. The myelin, including phospholipids, will be stained blue to green, and the neurons will
Catalog No. NC0098756