Learn More
MSC Luxor Shelf Utility Cart, 24 in Length, 18 in Width, Plastic, 3 Shelves
Catalog No. 5000532596
Encompass
The Luxor Shelf Utility Cart is a versatile and durable solution for material handling and storage. Constructed from plastic, this cart features three spacious shelves, making it ideal for various industrial and commercial settings. It measures 24 in length, 18 in width, and 35 in height, providing ample capacity for equipment and supplies.
  • Plastic construction ensures durability and ease of cleaning.
  • Three shelves provide generous storage and transport capacity.
  • Overall dimensions of 24 in length, 18 in width, and 35 in height.
  • Equipped with 4 in diameter wheels for smooth mobility.
  • Offers 11.75 in of clearance between shelves.
  • Lipped shelf profile helps secure items during transport.
  • Available in a sleek black finish.
  • Requires assembly.
Catalog No. 50-005-32596 Supplier MSC Supplier No. 33852732
The Luxor Shelf Utility Cart is a versatile and durable solution for material handling and storage. Constructed from plastic, this cart features three spacious shelves, making it ideal for various industrial and commercial settings. It measures 24 in length, 18 in width, and 35 in height, providing ample capacity for equipment and supplies.
  • Plastic construction ensures durability and ease of cleaning.
  • Three shelves provide generous storage and transport capacity.
  • Overall dimensions of 24 in length, 18 in width, and 35 in height.
  • Equipped with 4 in diameter wheels for smooth mobility.
  • Offers 11.75 in of clearance between shelves.
  • Lipped shelf profile helps secure items during transport.
  • Available in a sleek black finish.
  • Requires assembly.