Learn More
MONOD BIO INC LuxSit™ Pro Control Plasmid
Catalog No. NC3199873
Encompass_Preferred
The LuxSit™ Pro Control Plasmid encodes the LuxSit™ Pro enzyme under a CMV promoter for use with the LuxSit™ Pro Luciferase Assay System. The plasmid backbone is pcDNA3.1-CMV and is shipped lyophilized in 25 µg aliquots.The Monod Bio LuxSit™ Pro family features the world’s smallest and highest-performance de novo luciferases. Available as full-length reporters or split complementation systems, LuxSit™ Pro (13.5 kDa) is an ATP-independent luciferase designed with state-of-the-art machine learning. The enzyme is monomeric, soluble, thermostable, and expressible in both bacterial and mammalian cells. Combined with the LuxSit™ Pro Assay Buffer and Substrate, it produces glow-type luminescence several times brighter than Firefly or Renilla luciferases.Documentation and datasheets: monod.bio/access-luxsit-proLearn more at monod.bio
Catalog No. NC3199873 Supplier MONOD BIO INC Supplier No. P00103
The LuxSit™ Pro Control Plasmid encodes the LuxSit™ Pro enzyme under a CMV promoter for use with the LuxSit™ Pro Luciferase Assay System. The plasmid backbone is pcDNA3.1-CMV and is shipped lyophilized in 25 µg aliquots.The Monod Bio LuxSit™ Pro family features the world’s smallest and highest-performance de novo luciferases. Available as full-length reporters or split complementation systems, LuxSit™ Pro (13.5 kDa) is an ATP-independent luciferase designed with state-of-the-art machine learning. The enzyme is monomeric, soluble, thermostable, and expressible in both bacterial and mammalian cells. Combined with the LuxSit™ Pro Assay Buffer and Substrate, it produces glow-type luminescence several times brighter than Firefly or Renilla luciferases.Documentation and datasheets: monod.bio/access-luxsit-proLearn more at monod.bio