Learn More
MONOD BIO INC LuxSit™ Pro Plasmid, C-term
Catalog No. NC3199872
Encompass
The LuxSit™ Pro, C-term plasmid encodes the LuxSit™ Pro enzyme as a C-terminal reported tag for a protein of interest to be used with the LuxSit™ Pro Luciferase Assay System. The plasmid backbone is pcDNA3.1 with CMV as promoter. The gene of interest can be inserted in frame with the LuxSit™ Pro enzyme using (EcoRV, BstXI, XbaI, SacI, EcoRI, XhoI) restriction enzymes. The plasmid is shipped lyophilized in 25 ug aliquots.
Catalog No. NC3199872 Supplier MONOD BIO INC Supplier No. P00102
The LuxSit™ Pro, C-term plasmid encodes the LuxSit™ Pro enzyme as a C-terminal reported tag for a protein of interest to be used with the LuxSit™ Pro Luciferase Assay System. The plasmid backbone is pcDNA3.1 with CMV as promoter. The gene of interest can be inserted in frame with the LuxSit™ Pro enzyme using (EcoRV, BstXI, XbaI, SacI, EcoRI, XhoI) restriction enzymes. The plasmid is shipped lyophilized in 25 ug aliquots.