Learn More
Description
- Refreshing cranberry fragrance
- SANITARY SEALED™ refill helps prevent contamination
- ECOLOGO™ Certified to Standard Name UL 2784
- USDA Certified Biobased Product
- Fresh dispensing valve with each refill
Specifications
Specifications
| Product Type | Hand Cleanser |
| Color | Pink |
| Form | Foam |
| For Use With (Equipment) | GOJO™ FMX-20™ Dispenser |
| Quantity | 2/Pk. |
| Size | 2000 mL |
| Volume (Metric) | 2000 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

