Learn More
Medchemexpress LLC HY-101254 1mg Medchemexpress, Luzindole CAS:117946-91-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1012541MG
Encompass_Preferred
Medchemexpress, HY-101254 1mg Luzindole CAS:117946-91-5 Luzindole (N-0774) is a selective melatonin receptor antagonist. Luzindole preferentially targets MT2 (Mel1b) over MT1 (Mel1a) with Ki values of 10.2 and 158 nM for human MT2 and MT1, respectively. Luzindole suppresses experimental autoimmune encephalomyelitis (EAE), and exerts antidepressant-like activity[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7817