Learn More
Medchemexpress LLC HY-112711 10mg Medchemexpress, LV-320 CAS: Purity:>98%
SDP
Catalog No. 501969150
Encompass_Preferred
Medchemexpress, HY-112711 10mg LV-320 CAS: LV-320 is a potent and uncompetitive ATG4B inhibitor with an IC50 of 24.5 µM and a Kd of 16 µM. LV-320 inhibits ATG4B enzymatic activity, blocks autophagic flux in cells, and is stable, non-toxic and active in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9150 Supplier Medchemexpress LLC Supplier No. HY11271110MG
Medchemexpress, HY-112711 10mg LV-320 CAS: LV-320 is a potent and uncompetitive ATG4B inhibitor with an IC50 of 24.5 µM and a Kd of 16 µM. LV-320 inhibits ATG4B enzymatic activity, blocks autophagic flux in cells, and is stable, non-toxic and active in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.