Learn More
Medchemexpress LLC LV-320 | 2449093-46-1 | 95.0% | 520.11 | 1 ML
SDP
Catalog No. 5000437456
Encompass_Preferred
LV-320 is a potent and uncompetitive ATG4B inhibitor with an IC50 of 24.5 μM and a Kd of 16 μM. It inhibits ATG4B enzymatic activity, blocks autophagic flux in cells, and is stable, non-toxic, and active in vivo.
  • Increases endogenous LC3B-II and protein p62 levels in SKBR3, MCF7, JIMT1, and MDA-MB-231 cells in a dose-dependent manner.
  • Blocks autophagic flux in MDA-MB-231 cells.
  • Achieves terminal blood levels of 169 μM and liver levels of 104 μM in GFP-LC3 mice following oral gavage.
  • Significantly increases the accumulation of GFP-LC3 puncta and LC3B-II protein in treated animals without causing significant toxicity at tested doses.
Catalog No. 50-004-37456 Supplier Medchemexpress LLC Supplier No. HY11271110MM/1ML
LV-320 is a potent and uncompetitive ATG4B inhibitor with an IC50 of 24.5 μM and a Kd of 16 μM. It inhibits ATG4B enzymatic activity, blocks autophagic flux in cells, and is stable, non-toxic, and active in vivo.
  • Increases endogenous LC3B-II and protein p62 levels in SKBR3, MCF7, JIMT1, and MDA-MB-231 cells in a dose-dependent manner.
  • Blocks autophagic flux in MDA-MB-231 cells.
  • Achieves terminal blood levels of 169 μM and liver levels of 104 μM in GFP-LC3 mice following oral gavage.
  • Significantly increases the accumulation of GFP-LC3 puncta and LC3B-II protein in treated animals without causing significant toxicity at tested doses.