Learn More
Medchemexpress LLC HY-112711 5mg Medchemexpress, LV-320 CAS: Purity:>98%
SDP
Catalog No. 501932433
Encompass_Preferred
Medchemexpress, HY-112711 5mg LV-320 CAS: LV-320 is a potent and uncompetitive ATG4B inhibitor with an IC 50 of 24.5 µM and a K d of 16 µM. LV-320 inhibits ATG4B enzymatic activity, blocks autophagic flux in cells, and is stable, non-toxic and active in vivo [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2433 Supplier Medchemexpress LLC Supplier No. HY1127115MG
Medchemexpress, HY-112711 5mg LV-320 CAS: LV-320 is a potent and uncompetitive ATG4B inhibitor with an IC 50 of 24.5 µM and a K d of 16 µM. LV-320 inhibits ATG4B enzymatic activity, blocks autophagic flux in cells, and is stable, non-toxic and active in vivo [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.