Learn More
SignaGen Laboratories LV-EF1α-GFP-T2A-fLuc-Puro, Unit Size: 2 x 25 µL (>10^9 TU/mL)
Catalog No. NC3786871
Encompass
LV-EF1α-GFP-T2A-fLuc-Puro is a pre-made lentivirus that over-expresses eGFP and firefly luciferase (fLuc) via T2A linker under the EF1α promoter with co-expression of puromycin selection in an IRES cassette. Ready to use format.
Catalog No. NC3786871 Supplier SignaGen Laboratories Supplier No. SL100329
LV-EF1α-GFP-T2A-fLuc-Puro is a pre-made lentivirus that over-expresses eGFP and firefly luciferase (fLuc) via T2A linker under the EF1α promoter with co-expression of puromycin selection in an IRES cassette. Ready to use format.