Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LV-MAX™ Lentiviral Packaging Mix

Catalog No. A43237
Encompass_Preferred
Change view
Click to view available options
Quantity:
1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A43237 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A43237 Supplier Invitrogen™ Supplier No. A43237
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

LV-MAX Lentiviral Packaging Mix is an optimized single formulation containing a defined mixture of three packaging plasmids: pLP1, pLP2, and pLP/VSVG.

The LV-MAX Lentiviral Packaging Mix is a single, optimized set of packaging plasmids, designed to streamline and enhance the production of lentiviral vectors. This mix contains a defined combination of three essential packaging plasmids: pLP1, pLP2, and pLP/VSVG, which together supply the necessary structural and replication proteins in trans when expressed in HEK293-derived Viral Production Cells. This third-generation packaging system offers enhanced biosafety, as the Rev gene is supplied on a separate plasmid, independent of other lentiviral genes, thereby significantly reducing the likelihood of recombination and producing replication-competent lentiviral particles.

Key features of the LV-MAX Lentiviral Packaging Mix

  • Optimized formulation—combines three packaging plasmids (pLP1, pLP2, and pLP/VSVG) into a single mix, saving process development time by minimizing the need to optimize plasmid ratios
  • High titer—achieves high titers of lentiviral vectors when used with the LV-MAX Lentiviral Production System

Applications

  • Gene therapy—ideal for the production of lentiviral vectors for gene therapy research and development
  • Cell therapy—supports the generation of lentiviral vectors for cell therapy applications, facilitating seamless transition from bench to clinic
  • Research and development—provides a reliable and scalable solution for viral vector production in various research settings
TRUSTED_SUSTAINABILITY

Specifications

Product Type Lentiviral Packaging Mix
Sterility Non-sterile
Quantity 1.5 mL
Content And Storage Store at -5 to -30°C. Ships on gel ice.

Are the LV-MAX Lentiviral Packaging Mix (Cat. No. A43237) and ViraPower Lentiviral Packaging Mix (Cat. No. K497500) interchangeable?

Yes, they can be used interchangeably.

What are the major differences between LV-MAX and CTS LV- MAX products?

The LV-MAX and CTS LV- MAX products have equivalent performance. In contrast to the research use only LV-MAX products, CTS LV-MAX products have an additional intended use for cell and gene therapy applications, and are complemented with a more robust regulatory support package to enable clinical therapeutic applications. This means that to the best of our ability, we keep our CTS product specifications and release requirements up-to-date in accordance with the latest regulatory guidance specific for this intended use.

What is the intended use claim for your LV-MAX and CTS LV-MAX products?

Our LV-MAX products are intended for research use only while our CTS LV- MAX products are intended for research use or manufacture of cell, gene or tissue based products. Please refer to the individual products for further details on the intended use claim.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.