Learn More
Brookfield Engineer Labs LV SET (4 SPINDLES)
Catalog No. NC9618033
Encompass
LV Standard Threaded Spindle Set of four spindles (61, 62, 63, 64)
Catalog No. NC9618033 Supplier Brookfield Engineer Labs Supplier No. SSL
LV Standard Threaded Spindle Set of four spindles (61, 62, 63, 64)