Learn More
Brookfield Engineer Labs LV SPINDLE LV-2LV SPINDLE LV
Catalog No. NC9144044
Encompass
Spindle, Entry Code: 62, SMC: 32, SRC: 0, Each spindle has a two digit entry code which is entered via the keypad on the DV-II +, The entry codeallows the DV-II + to calculate Viscosity, Shear Rate and Shear Stress values.
Catalog No. NC9144044 Supplier Brookfield Engineer Labs Supplier No. LV2
Spindle, Entry Code: 62, SMC: 32, SRC: 0, Each spindle has a two digit entry code which is entered via the keypad on the DV-II +, The entry codeallows the DV-II + to calculate Viscosity, Shear Rate and Shear Stress values.