Learn More
Takara Bio Living Colors ZsGreen Monoclonal Antibody | 100 uL
SDP
Supplier:  Takara Bio 632598
Encompass
The Living Colors ZsGreen Monoclonal Antibody was raised in mouse against recombinant full-length Zoanthus sp. green fluorescent protein (ZsGreen), and Protein A-purified. In Western blot applications, it specifically recognizes ZsGreen and does not cross-react with AcGFP1, AmCyan, ZsYellow, DsRed-Express, mCherry, tdTomato, Dendra2, or DsRed2.
Catalog No. NC1252852