Learn More
Cayman Chemical L-Valacyclovir (hydrochloride), Purity: ≥98%, CAS: 124832-27-5, 50 mg
SDP
Catalog No. 502964466
Encompass_Preferred
A prodrug of acyclovir; inhibits HSV-I replication (IC50 = 0.84 µM); rapidly converted to acyclovir in vivo; active against varicella-zoster and Epstein-Barr viruses in vitro (IC50s = 1.3-20.6 and 0.3-25 µM, respectively)
Catalog No. 50-296-4466 Supplier Cayman Chemical Supplier No. 2380150 MG
A prodrug of acyclovir; inhibits HSV-I replication (IC50 = 0.84 µM); rapidly converted to acyclovir in vivo; active against varicella-zoster and Epstein-Barr viruses in vitro (IC50s = 1.3-20.6 and 0.3-25 µM, respectively)