Learn More
MSC HYFLEX C/R POLY GLV
Catalog No. 502738566
Encompass
Cut-Resistant Gloves: Size Medium, ANSI Cut A2, ANSI Puncture 2, Polyurethane, Series 11-518
Catalog No. 50-273-8566 Supplier MSC Supplier No. 51611978
Add to Cart
Add to Cart
Cut-Resistant Gloves: Size Medium, ANSI Cut A2, ANSI Puncture 2, Polyurethane, Series 11-518