Learn More
MSC ALPHATEC APOLLO SUIT
Catalog No. 502736656
Encompass
Rain & Chemical Wear; Size: 2X-Large ; Color: Orange ; Material: Multi-Layer Non-Woven Barrier Laminate Fabric ; Garment Type: Chemical-Resistant ; High Visibility: Yes ; Hood Type: Attached Hood
Catalog No. 50-273-6656 Supplier MSC Supplier No. 86713666
Add to Cart
Add to Cart
Rain & Chemical Wear; Size: 2X-Large ; Color: Orange ; Material: Multi-Layer Non-Woven Barrier Laminate Fabric ; Garment Type: Chemical-Resistant ; High Visibility: Yes ; Hood Type: Attached Hood