Learn More
Leybold Vacuum Products Inc Leybonol LVO 210 Synthetic Oil, 5 L
Catalog No. NC1533226
Encompass_Preferred
Synthetic oil (ester oil with additives). Very high thermal, oxidative and chemical stability, good deterging and dispersion characteristics, excellent wear protection. For high temperature applications, pumping of air, chemically inert gases, carbon dioxide (dry),carbon monoxide, organic solvent vapors, resin vapors, etc.
Catalog No. NC1533226 Supplier Leybold Vacuum Products Inc Supplier No. L21005
Synthetic oil (ester oil with additives). Very high thermal, oxidative and chemical stability, good deterging and dispersion characteristics, excellent wear protection. For high temperature applications, pumping of air, chemically inert gases, carbon dioxide (dry),carbon monoxide, organic solvent vapors, resin vapors, etc.