Learn More
Aviva Systems Biology Corp LW-1 Antibody - middle region (ARP35905_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35905_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LW-1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human LW-1. Alternate names: LW-1, HSFX2ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-55918