Learn More
LW Scientific Digital Differential Counters
Catalog No. 22500102 Shop All LW Scientific Products
Change view
Click to view available options
No. of Keys:
Ten buttons
Catalog No. No. of Keys
22-500-102 Ten buttons
1 options
Catalog No. 22-500-102 Supplier LW Scientific Supplier No. CTLDIFD10K3
  • Digital readout with soft-touch buttons
  • Electronic with LED display
  • Ten buttons for counting with audible beeps
  • Audible long beep when 100 cells are counted
  • No mechanical gears
  • Ten buttons–8 named cells + 2 blanks
  • Pictures and names of blood cells next to each button
  • Ten 2-digit windows + one total 3-digit window
  • Reset button to clear all counts
  • Percentage button for any-time percentage calculations
  • 110V-220V Auto-switching
Specifications
No. of Keys Ten buttons
Weight (Metric) 0.58kg
Voltage 110V-220V Auto-switching