Learn More
World Precision Instrument LWCC Calibration Solution
SDP
Catalog No. 502538727
Encompass
  • This solution is the one used in WPI laboratories.
  • The solution is used to replicate the specification on the quality control sheet that is shipped with flow cells to verify values before starting experiments.
  • Good practice to calibrate flow cells after they have been stored.
  • Volume of 125 mL
  • Package of 5
Catalog No. 50-253-8727 Supplier World Precision Instrument Supplier No. LWCCCAL
  • This solution is the one used in WPI laboratories.
  • The solution is used to replicate the specification on the quality control sheet that is shipped with flow cells to verify values before starting experiments.
  • Good practice to calibrate flow cells after they have been stored.
  • Volume of 125 mL
  • Package of 5