Learn More
World Precision Instrument LWCC Start-up Kit
SDP
Catalog No. 502538734
Encompass
Startup kit for LWCC liquid waveguide capillary cells includes 2 WVLUXUVIS-S-600 Fiber Optic cables (1 meter, SMA cconnector, 600 mm core, UV enhanced), sample injector assembly, Ministar Peristaltic Pump with 6 rollers for smooth liquid injection, and 501609 Wavegide Cleaning Kit (commonly used solutions for cleaning LWCC)
Catalog No. 50-253-8734 Supplier World Precision Instrument Supplier No. KITLWCC
Startup kit for LWCC liquid waveguide capillary cells includes 2 WVLUXUVIS-S-600 Fiber Optic cables (1 meter, SMA cconnector, 600 mm core, UV enhanced), sample injector assembly, Ministar Peristaltic Pump with 6 rollers for smooth liquid injection, and 501609 Wavegide Cleaning Kit (commonly used solutions for cleaning LWCC)