Learn More
World Precision Instrument LWS Professional Grade Binocular Microscope
SDP
Catalog No. 502538402
Encompass
Best-seller in universities, medical schools, research laboratories
The W30S professional-grade microscope is a best-seller in universities, medical schools, and research laboratories. Equipped for performance, its features include titanium-finished DIN or Semi-Plan optics and a 30-year antifungal coating. The W30S is the choice for superior performance at a great price.
Catalog No. 50-253-8402 Supplier World Precision Instrument Supplier No. W30SLED
Best-seller in universities, medical schools, research laboratories
The W30S professional-grade microscope is a best-seller in universities, medical schools, and research laboratories. Equipped for performance, its features include titanium-finished DIN or Semi-Plan optics and a 30-year antifungal coating. The W30S is the choice for superior performance at a great price.